Assay Details
| Gene Symbol: Sars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023637.3 | NP_076126.2 | 13 - 14 | 1303 | 54 | |
| GenBank mRNA | AB029949.1 | - | 13 - 14 | 1275 | 54 | |
| AK010491.1 | - | 13 - 14 | 1281 | 54 | ||
| AK087606.1 | - | 13 - 14 | 1270 | 54 | ||
| AK140760.1 | - | 13 - 14 | 1270 | 54 | ||
| BC079664.1 | - | 13 - 14 | 1280 | 30357834 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sars2
Genomic Map