Assay Details
| Gene Symbol: Pank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114339.2 | NP_001107811.1 | 4 - 5 | 1713 | 85 | |
| NM_023792.2 | NP_076281.1 | 4 - 5 | 1013 | 85 | ||
| GenBank mRNA | AF200357.1 | - | 4 - 5 | 1013 | 85 | |
| AF347700.1 | - | 4 - 5 | 1711 | 85 | ||
| AK017345.1 | - | 4 - 5 | 1580 | 85 | ||
| AK050222.1 | - | 4 - 5 | 1014 | 85 | ||
| AK184272.1 | - | 2 - 3 | 203 | 85 | ||
| BC023496.1 | - | 4 - 5 | 935 | 5054239 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pank1
Genomic Map