Assay Details
| Gene Symbol: Sapcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023893.4 | NP_076382.3 | 2 - 3 | 361 | 124 | |
| NR_028258.1 | 2 - 3 | 365 | 124 | |||
| GenBank mRNA | AK019275.1 | - | 2 - 3 | 262 | 124 | |
| BC147772.1 | - | 2 - 3 | 361 | 9087495 | 124 | |
| BC147780.1 | - | 2 - 3 | 361 | 9087500 | 124 | |
| BC172157.1 | - | 2 - 3 | 124 | |||
| CF739520.1 | - | 2 - 3 | 259 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637935 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140091 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157125 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140091 | - | - |
| Whole Mouse Genome G4122A | A_52_P70064 | BC172157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70064 | BF465693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70064 | CF739520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70064 | NM_023893.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P70064 | NR_028258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | BC172157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | BF465693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | CF739520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | NM_023893.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233727 | NR_028258.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157125 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450380 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422952_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450379 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444637 | - | - |
| Mouse Genome 430 2.0 Array | 1422952_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422952_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78376_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78376_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018373.mm.1 | NM_023893.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018373.mm.1 | NM_023893.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018373.mm.1 | TC1700001939.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018375.mm.1 | NM_023893.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018375.mm.1 | NM_023893.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018375.mm.1 | NR_028258.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018375.mm.1 | NR_028258.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018375.mm.1 | TC1700001939.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018370.mm.1 | NM_023893.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018370.mm.1 | NM_023893.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018370.mm.1 | NR_028258.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018370.mm.1 | NR_028258.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018370.mm.1 | TC1700001939.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018374.mm.1 | NM_023893.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018374.mm.1 | NM_023893.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018374.mm.1 | NR_028258.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018374.mm.1 | NR_028258.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018374.mm.1 | TC1700001939.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018371.mm.1 | NM_023893.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018371.mm.1 | NM_023893.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018371.mm.1 | TC1700001939.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422952_at | - | - |
| Mouse Expression Set 430 | 1422952_PM_at | - | - |
| Murine Genome U74v2 Set | 92307_at | - | - |
| Murine Genome U74v2 Set | 92307_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422952_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422952_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422952_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422952_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 422022 | - | - |
| ABI Mouse Genome Survey Microarray | 519306 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6400400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239632 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234510 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234510 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239632 | - | - |

Sapcd1
Genomic Map