Assay Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024193.2 | NP_077155.2 | 2 - 3 | 156 | 70 | |
| GenBank mRNA | AK009799.1 | - | 2 - 3 | 142 | 70 | |
| AK013528.1 | - | 2 - 3 | 123 | 70 | ||
| AK030730.1 | - | 2 - 3 | 125 | 70 | ||
| AK077795.1 | - | 2 - 3 | 123 | 70 | ||
| AK139924.1 | - | 2 - 3 | 123 | 70 | ||
| AK146243.1 | - | 2 - 3 | 123 | 70 | ||
| AK150258.1 | - | 2 - 3 | 123 | 70 | ||
| AK154024.1 | - | 2 - 3 | 124 | 70 | ||
| BC021355.1 | - | 2 - 3 | 118 | 5006404 | 70 | |
| CJ162437.1 | - | 2 - 3 | 159 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nop56
Genomic Map