Assay Details
| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024245.4 | NP_077207.3 | 3 - 4 | 380 | 68 | |
| GenBank mRNA | AK013939.1 | - | 3 - 4 | 318 | 68 | |
| AK030771.1 | - | 3 - 4 | 357 | 68 | ||
| BC003363.1 | - | 3 - 4 | 353 | 68 | ||
| BC047273.1 | - | 3 - 4 | 338 | 6440797 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif23
Genomic Map