Assay Details
| Gene Symbol: Akr7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025337.3 | NP_079613.3 | 5 - 6 | 824 | 60 | |
| GenBank mRNA | AF525358.2 | - | 5 - 6 | 726 | 60 | |
| AJ271800.1 | - | 5 - 6 | 741 | 60 | ||
| AJ271801.1 | - | 5 - 6 | 741 | 60 | ||
| AK002664.1 | - | 5 - 6 | 804 | 60 | ||
| AK003915.1 | - | 5 - 6 | 811 | 60 | ||
| AK143203.1 | - | 5 - 6 | 823 | 60 | ||
| BC031857.1 | - | 5 - 6 | 801 | 5124841 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351044 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351040 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770851 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164675 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23351044 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164675 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05424738 | - | - |
| Whole Mouse Genome G4122A | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | AK002664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | AK003915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | AK143203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | BC031857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188795 | NM_025337.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123778 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164675 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03481822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03481806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03481804 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351044 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23351043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23351047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03481815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04053532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04053527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03481810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23351083 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509636 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438315_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417294_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458300_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509635 | - | - |
| Mouse Genome 430 2.0 Array | 1438315_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1458300_at | - | - |
| Mouse Genome 430 2.0 Array | 1438315_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417294_at | - | - |
| Mouse Genome 430 2.0 Array | 1458300_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417294_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110198_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110198_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400014265.mm.1 | NM_025337.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014265.mm.1 | NM_025337.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014265.mm.1 | TC0400001743.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438315_x_at | - | - |
| Mouse Expression Set 430 | 1417294_at | - | - |
| Mouse Expression Set 430 | 1417294_PM_at | - | - |
| Mouse Expression Set 430 | 1438315_PM_x_at | - | - |
| Mouse Expression Set 430 | 1458300_at | - | - |
| Mouse Expression Set 430 | 1458300_PM_at | - | - |
| Murine Genome U74v2 Set | 96069_PM_at | - | - |
| Murine Genome U74v2 Set | 96069_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438315_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417294_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417294_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438315_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417294_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417294_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438315_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438315_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458300_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458300_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260341 | - | - |
| MouseWG-6 v2 Expression BeadChip | 460672 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 460672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625991 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256518 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256518 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2625991 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002846.1_27-S | - | - |
| mouse-6 v1.1 (Illumina) | scl24777.7.1_203-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460672 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002846.1_27-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24777.7.1_203-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2908045 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2625991 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256518 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256518 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625991 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2908045 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24777.7.1_203-S | - | - |

Akr7a5
Genomic Map