Assay Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025638.2 | NP_079914.1 | 6 - 7 | 740 | 71 | |
| GenBank mRNA | AK011487.1 | - | 6 - 7 | 663 | 71 | |
| AK013864.1 | - | 6 - 7 | 666 | 71 | ||
| AK016023.1 | - | 6 - 7 | 666 | 71 | ||
| AK167812.1 | - | 6 - 7 | 723 | 71 | ||
| BC016541.1 | - | 6 - 7 | 663 | 3602449 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd1
Genomic Map