Assay Details
| Gene Symbol: Sltm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025690.3 | NP_079966.2 | 17 - 18 | 2417 | 79 | |
| NM_026337.1 | NP_080613.1 | 17 - 18 | 2455 | 79 | ||
| GenBank mRNA | AF462146.1 | - | 17 - 18 | 2417 | 79 | |
| AK134756.1 | - | 8 - 9 | 1096 | 79 | ||
| BC019992.1 | - | 6 - 7 | 787 | 79 | ||
| BC139202.1 | - | 17 - 18 | 2407 | 8862224 | 79 | |
| BC145577.1 | - | 16 - 17 | 2226 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sltm
Genomic Map