| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300011820.mm.1 |
NM_025772.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011820.mm.1 |
NM_025772.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011820.mm.1 |
TC1300001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011817.mm.1 |
NM_025772.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011817.mm.1 |
NM_025772.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011817.mm.1 |
TC1300001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011815.mm.1 |
NM_025772.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011815.mm.1 |
NM_025772.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011815.mm.1 |
TC1300001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011818.mm.1 |
NM_025772.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011818.mm.1 |
NM_025772.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011818.mm.1 |
TC1300001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011819.mm.1 |
NM_025772.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011819.mm.1 |
NM_025772.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011819.mm.1 |
TC1300001902.mm.1 |
MTA TC |