Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 7 - 8 | 631 | 81 | |
| GenBank mRNA | AJ404859.1 | - | 7 - 8 | 630 | 81 | |
| AK010924.1 | - | 3 - 4 | 243 | 81 | ||
| AK017400.1 | - | 3 - 4 | 409 | 81 | ||
| AK159656.1 | - | 7 - 8 | 756 | 81 | ||
| AY265461.1 | - | 3 - 4 | 409 | 81 | ||
| BC018350.1 | - | 7 - 8 | 622 | 4195234 | 81 | |
| BC048682.1 | - | 7 - 8 | 676 | 81 | ||
| BC058574.1 | - | 7 - 8 | 604 | 6487720 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dtnbp1
Genomic Map