Assay Details
| Gene Symbol: Wwp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025830.3 | NP_080106.1 | 18 - 19 | 2032 | 81 | |
| GenBank mRNA | AK004962.2 | - | 18 - 19 | 2048 | 81 | |
| AK088936.1 | - | 18 - 19 | 2011 | 81 | ||
| AK146306.1 | - | 17 - 18 | 1894 | 81 | ||
| AK151548.1 | - | 18 - 19 | 2026 | 81 | ||
| AK159248.1 | - | 9 - 10 | 1076 | 81 | ||
| AK181554.1 | - | 1 - 2 | 77 | 81 | ||
| BC004712.1 | - | 3 - 4 | 352 | 81 | ||
| BC039921.1 | - | 18 - 19 | 2024 | 4954925 | 81 | |
| BC048184.1 | - | 18 - 19 | 2032 | 6827381 | 81 | |
| HQ896246.1 | - | 9 - 10 | 1129 | 81 | ||
| JN712751.1 | - | 18 - 19 | 2065 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wwp2
Genomic Map