Assay Details
| Gene Symbol: Wwp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025830.3 | NP_080106.1 | 20 - 21 | 2294 | 80 | |
| GenBank mRNA | AK004962.2 | - | 20 - 21 | 2310 | 80 | |
| AK088936.1 | - | 20 - 21 | 2273 | 80 | ||
| AK090392.1 | - | 144 | 80 | |||
| AK146306.1 | - | 19 - 20 | 2156 | 80 | ||
| AK151548.1 | - | 20 - 21 | 2288 | 80 | ||
| AK159248.1 | - | 11 - 12 | 1338 | 80 | ||
| BC004712.1 | - | 5 - 6 | 614 | 80 | ||
| BC018496.1 | - | 4 - 5 | 1492 | 80 | ||
| BC039921.1 | - | 20 - 21 | 2286 | 4954925 | 80 | |
| BC048184.1 | - | 20 - 21 | 2294 | 6827381 | 80 | |
| HQ896246.1 | - | 11 - 12 | 1391 | 80 | ||
| JN712751.1 | - | 20 - 21 | 2327 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wwp2
Genomic Map