Assay Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026257.3 | NP_080533.1 | 12 - 13 | 1168 | 74 | |
| NR_045571.1 | 11 - 12 | 1047 | 74 | |||
| GenBank mRNA | AK010492.1 | - | 5 - 6 | 647 | 74 | |
| AK015720.1 | - | 12 - 13 | 1152 | 74 | ||
| AK048494.1 | - | 12 - 13 | 1170 | 74 | ||
| BC019461.1 | - | 11 - 12 | 1024 | 4224033 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn11
Genomic Map