Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 3 - 4 | 474 | 94 | |
| GenBank mRNA | AK003264.1 | - | 3 - 4 | 474 | 94 | |
| AK017161.1 | - | 2 - 3 | 220 | 94 | ||
| AK018002.1 | - | 3 - 4 | 471 | 94 | ||
| AK132000.1 | - | 3 - 4 | 475 | 94 | ||
| AK146710.1 | - | 3 - 4 | 476 | 94 | ||
| BC036284.1 | - | 3 - 4 | 482 | 5136304 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat8
Genomic Map