Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 8 - 9 | 1019 | 68 | |
| GenBank mRNA | AK003264.1 | - | 8 - 9 | 1019 | 68 | |
| AK008418.1 | - | 8 - 9 | 1006 | 68 | ||
| AK017161.1 | - | 7 - 8 | 765 | 68 | ||
| AK018002.1 | - | 8 - 9 | 1015 | 68 | ||
| AK132000.1 | - | 8 - 9 | 1020 | 68 | ||
| AK146710.1 | - | 8 - 9 | 1021 | 68 | ||
| BC036284.1 | - | 8 - 9 | 1027 | 5136304 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat8
Genomic Map