Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026393.2 | NP_080669.1 | 2 - 3 | 706 | 62 | |
| GenBank mRNA | BC030039.1 | - | 2 - 3 | 409 | 1313824 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Nmral1
Genomic Map