Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026393.2 | NP_080669.1 | 3 - 4 | 731 | 68 | |
| GenBank mRNA | BC030039.1 | - | 3 - 4 | 434 | 1313824 | 68 |
More Information
Set Membership: |
|

Nmral1
Genomic Map