Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290761.1 | NP_001277690.1 | 4 - 5 | 1141 | 92 | |
| NM_026393.2 | NP_080669.1 | 5 - 6 | 1171 | 92 | ||
| GenBank mRNA | AK146521.1 | - | 897 | 92 | ||
| BC030039.1 | - | 5 - 6 | 874 | 1313824 | 92 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511777 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511776 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137521 | BC030039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137521 | NM_026393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137521 | AK012340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137521 | AK078389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137521 | AK146521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337425 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179808 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137521 | BC030039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137521 | NM_026393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137521 | AK012340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137521 | AK078389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137521 | AK146521.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30511765 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08493148 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179808 | - | - |
| Whole Mouse Genome G4122A | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | BC030039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | NM_026393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | AK012340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480233 | AK146521.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123463 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12968879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12764527 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12693326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13021911 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12764526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13021909 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13021910 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511776 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511777 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30511779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30511737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12666591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12666594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12764527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12741797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12902014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12902015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12588418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12666590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12591213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12484041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12905096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12780790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12522552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12968879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12666592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13053388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13021911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12741793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30511747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437442 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430530_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453562_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453563_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437432 | - | - |
| Mouse Genome 430 2.0 Array | 1430530_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1430530_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453563_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453563_at | - | - |
| Mouse Genome 430 2.0 Array | 1453562_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453562_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67824_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67824_PM_at | - | - |
| Mouse Expression Set 430 | 1430530_s_at | - | - |
| Mouse Expression Set 430 | 1453562_a_at | - | - |
| Mouse Expression Set 430 | 1430530_PM_s_at | - | - |
| Mouse Expression Set 430 | 1453563_at | - | - |
| Mouse Expression Set 430 | 1453563_PM_at | - | - |
| Mouse Expression Set 430 | 1453562_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453563_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453562_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430530_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453562_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430530_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453563_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 919849 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106040484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106040484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4210239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232252 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239181 | - | - |
| mouse-6 v1.1 (Illumina) | scl48804.7.78_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106040484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48804.7.78_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106040484 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239181 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232252 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl48804.7.78_52-S | - | - |

Nmral1
Genomic Map