Assay Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026924.3 | NP_081200.2 | 1 - 2 | 318 | 59 | |
| GenBank mRNA | AB101294.1 | - | 1 - 2 | 221 | 59 | |
| AK167697.1 | - | 1 - 2 | 309 | 59 | ||
| AY090537.1 | - | 1 - 2 | 129 | 59 | ||
| BC031771.1 | - | 1 - 2 | 318 | 4219296 | 59 | |
| BC057210.1 | - | 1 - 2 | 339 | 5329625 | 59 | |
| BC094445.1 | - | 1 - 2 | 330 | 4981833 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ovol2
Genomic Map