Assay Details
| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027052.3 | NP_081328.2 | 10 - 11 | 1115 | 70 | |
| GenBank mRNA | AB055004.1 | - | 10 - 11 | 1091 | 70 | |
| AK051023.1 | - | 10 - 11 | 1116 | 70 | ||
| AK164292.1 | - | 10 - 11 | 1058 | 70 | ||
| AK167539.1 | - | 10 - 11 | 1097 | 70 | ||
| AK169349.1 | - | 9 - 10 | 1104 | 70 | ||
| AY027919.1 | - | 10 - 11 | 1154 | 70 | ||
| BC024072.1 | - | 10 - 11 | 1040 | 5097293 | 70 | |
| BC024123.1 | - | 10 - 11 | 1072 | 5098067 | 70 | |
| BC031717.1 | - | 10 - 11 | 1033 | 4195569 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a4
Genomic Map