Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700037507.mm.1 |
NM_001045483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037507.mm.1 |
NM_001045483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037507.mm.1 |
NM_027115.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037507.mm.1 |
NM_027115.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037507.mm.1 |
TC0700004473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037503.mm.1 |
NM_001045483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037503.mm.1 |
NM_001045483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037503.mm.1 |
NM_027115.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037503.mm.1 |
NM_027115.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037503.mm.1 |
TC0700004473.mm.1 |
MTA TC |