Assay Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 6 - 7 | 682 | 85 | |
| GenBank mRNA | AK011847.1 | - | 6 - 7 | 652 | 85 | |
| AK173195.1 | - | 6 - 7 | 643 | 85 | ||
| BC017645.1 | - | 6 - 7 | 644 | 85 | ||
| BC038265.1 | - | 6 - 7 | 623 | 85 | ||
| BC137736.1 | - | 6 - 7 | 631 | 8860755 | 85 | |
| BC137737.1 | - | 6 - 7 | 631 | 8860756 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rufy2
Genomic Map