Assay Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 16 - 17 | 1702 | 100 | |
| GenBank mRNA | AK173195.1 | - | 16 - 17 | 1663 | 100 | |
| BC137736.1 | - | 16 - 17 | 1651 | 8860755 | 100 | |
| BC137737.1 | - | 16 - 17 | 1651 | 8860756 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rufy2
Genomic Map