Assay Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290390.1 | NP_001277319.1 | 19 - 20 | 3249 | 83 | |
| NM_001290392.1 | NP_001277321.1 | 16 - 17 | 2984 | 83 | ||
| NM_001290393.1 | NP_001277322.1 | 18 - 19 | 3198 | 83 | ||
| NM_027671.5 | NP_081947.2 | 19 - 20 | 3252 | 83 | ||
| GenBank mRNA | AF367759.1 | - | 18 - 19 | 1904 | 83 | |
| AK016927.1 | - | 5 - 6 | 541 | 83 | ||
| AK038698.1 | - | 16 - 17 | 2985 | 83 | ||
| AK080034.1 | - | 19 - 20 | 2614 | 83 | ||
| AK083241.1 | - | 19 - 20 | 2237 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sntg1
Genomic Map