Assay Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163746.1 | NP_001157218.1 | 7 - 8 | 1063 | 79 | |
| NM_028352.4 | NP_082628.3 | 7 - 8 | 1063 | 79 | ||
| GenBank mRNA | AK013402.1 | - | 7 - 8 | 1046 | 79 | |
| AK028066.1 | - | 7 - 8 | 1053 | 79 | ||
| AK032776.1 | - | 7 - 8 | 1015 | 79 | ||
| AK036561.1 | - | 7 - 8 | 1049 | 79 | ||
| AK043547.1 | - | 7 - 8 | 1064 | 79 | ||
| AK049337.1 | - | 7 - 8 | 1015 | 79 | ||
| AK049758.1 | - | 7 - 8 | 1050 | 79 | ||
| AK051706.1 | - | 7 - 8 | 1003 | 79 | ||
| AK153501.1 | - | 6 - 7 | 883 | 79 | ||
| AK160913.1 | - | 7 - 8 | 1064 | 79 | ||
| AK165513.1 | - | 7 - 8 | 1048 | 79 | ||
| AK169082.1 | - | 7 - 8 | 1028 | 79 | ||
| AK169141.1 | - | 7 - 8 | 1022 | 79 | ||
| AK169787.1 | - | 7 - 8 | 1037 | 79 | ||
| BC138700.1 | - | 7 - 8 | 988 | 8861722 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgm3
Genomic Map