Assay Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002238.2 | NP_001002238.1 | 10 - 11 | 1365 | 65 | |
| NM_001002240.2 | NP_001002240.1 | 10 - 11 | 1338 | 65 | ||
| NM_001002241.2 | NP_001002241.1 | 9 - 10 | 1638 | 65 | ||
| NM_031387.3 | NP_113564.2 | 10 - 11 | 1498 | 65 | ||
| GenBank mRNA | AB067571.1 | - | 9 - 10 | 1319 | 65 | |
| AB183526.1 | - | 10 - 11 | 1365 | 65 | ||
| AB183527.1 | - | 10 - 11 | 1338 | 65 | ||
| AB183528.1 | - | 9 - 10 | 1638 | 65 | ||
| AB183529.1 | - | 9 - 10 | 1202 | 65 | ||
| AF285591.1 | - | 6 - 7 | 692 | 65 | ||
| AK029806.1 | - | 10 - 11 | 1499 | 65 | ||
| BC129954.1 | - | 5 - 6 | 624 | 40081321 | 65 | |
| BC129955.1 | - | 5 - 6 | 624 | 40081323 | 65 | |
More Information
Set Membership: |
|

Tdrd1
Genomic Map