Assay Details
| Gene Symbol: Tnfrsf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291010.1 | NP_001277939.1 | 3 - 4 | 453 | 60 | |
| NM_033042.4 | NP_149031.2 | 2 - 3 | 514 | 60 | ||
| GenBank mRNA | AF329969.1 | - | 3 - 4 | 457 | 60 | |
| AK040632.1 | - | 2 - 3 | 422 | 60 | ||
| BC017526.1 | - | 2 - 3 | 514 | 4912933 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf25
Genomic Map