Assay Details
| Gene Symbol: Tnfrsf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291010.1 | NP_001277939.1 | 5 - 6 | 678 | 72 | |
| GenBank mRNA | AF329969.1 | - | 5 - 6 | 682 | 72 | |
More Information
Set Membership: |
|

Tnfrsf25
Genomic Map