| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100013455.mm.1 |
NM_001145800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013455.mm.1 |
NM_001145800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013455.mm.1 |
NM_033608.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013455.mm.1 |
NM_033608.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013455.mm.1 |
TC0100001616.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013456.mm.1 |
NM_001145800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013456.mm.1 |
NM_001145800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013456.mm.1 |
NM_033608.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013456.mm.1 |
NM_033608.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013456.mm.1 |
TC0100001616.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013457.mm.1 |
NM_001145800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013457.mm.1 |
NM_001145800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013457.mm.1 |
NM_033608.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013457.mm.1 |
NM_033608.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013457.mm.1 |
TC0100001616.mm.1 |
MTA TC |