Assay Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145800.1 | NP_001139272.1 | 19 - 20 | 3484 | 81 | |
| NM_033608.3 | NP_291086.2 | 19 - 20 | 3550 | 81 | ||
| GenBank mRNA | AF317839.1 | - | 19 - 20 | 3550 | 81 | |
| AK145121.1 | - | 19 - 20 | 3464 | 81 | ||
| BC023011.1 | - | 1 - 2 | 344 | 81 | ||
| BC024487.1 | - | 1 - 2 | 322 | 81 | ||
| BC034594.1 | - | 18 - 19 | 3313 | 81 | ||
| BC040281.1 | - | 19 - 20 | 3510 | 5364073 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf9
Genomic Map