Assay Details
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039533.2 | NP_001034622.1 | 8 - 9 | 939 | 62 | |
| NM_001291017.1 | NP_001277946.1 | 8 - 9 | 939 | 62 | ||
| NM_053181.3 | NP_444411.2 | 8 - 9 | 939 | 62 | ||
| GenBank mRNA | AK004611.1 | - | 8 - 9 | 816 | 62 | |
| AK030613.1 | - | 8 - 9 | 770 | 62 | ||
| AK033107.1 | - | 8 - 9 | 770 | 62 | ||
| AK054329.1 | - | 8 - 9 | 922 | 62 | ||
| AK075829.1 | - | 1 - 2 | 63 | 62 | ||
| AK163696.1 | - | 8 - 9 | 810 | 62 | ||
| AK165587.1 | - | 8 - 9 | 774 | 62 | ||
| AK170661.1 | - | 8 - 9 | 926 | 62 | ||
| BC005541.1 | - | 8 - 9 | 757 | 3590316 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK075829.1 transcripts.
Set Membership: |
|

Pdxdc1
Genomic Map