Assay Details
| Gene Symbol: Atp6v0a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080467.3 | NP_536715.3 | 12 - 13 | 1594 | 59 | |
| GenBank mRNA | AB050903.1 | - | 11 - 12 | 1320 | 59 | |
| AF326316.1 | - | 12 - 13 | 1388 | 59 | ||
| AF435090.1 | - | 12 - 13 | 1582 | 59 | ||
| AK128892.1 | - | 12 - 13 | 1594 | 59 | ||
| BC046979.1 | - | 13 - 14 | 1616 | 6315485 | 59 | |
More Information
Set Membership: |
|

Atp6v0a4
Genomic Map