Assay Details
| Gene Symbol: Wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080848.2 | NP_543124.1 | 3 - 4 | 405 | 87 | |
| GenBank mRNA | AF416510.1 | - | 3 - 4 | 410 | 87 | |
| AK075937.1 | - | 3 - 4 | 348 | 87 | ||
| AK144091.1 | - | 3 - 4 | 349 | 87 | ||
| BC016103.1 | - | 3 - 4 | 340 | 4919781 | 87 | |
| BC025801.1 | - | 3 - 4 | 382 | 5132525 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr5
Genomic Map