Assay Details
| Gene Symbol: Wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080848.2 | NP_543124.1 | 7 - 8 | 747 | 85 | |
| GenBank mRNA | AF416510.1 | - | 7 - 8 | 752 | 85 | |
| AK075937.1 | - | 7 - 8 | 690 | 85 | ||
| BC008547.1 | - | 2 - 3 | 127 | 85 | ||
| BC016103.1 | - | 7 - 8 | 682 | 4919781 | 85 | |
| BC025801.1 | - | 7 - 8 | 724 | 5132525 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr5
Genomic Map