Assay Details
| Gene Symbol: Xpnpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289729.1 | NP_001276658.1 | 10 - 11 | 1643 | 112 | |
| NM_133213.2 | NP_573476.2 | 10 - 11 | 1442 | 112 | ||
| GenBank mRNA | AF367247.1 | - | 10 - 11 | 1106 | 112 | |
| AF428102.1 | - | 10 - 11 | 1293 | 112 | ||
| AK078835.1 | - | 10 - 11 | 1249 | 112 | ||
| AK085520.1 | - | 10 - 11 | 1441 | 112 | ||
| AK087627.1 | - | 10 - 11 | 1249 | 112 | ||
| AK164538.1 | - | 10 - 11 | 1270 | 112 | ||
| AK172086.1 | - | 10 - 11 | 1443 | 112 | ||
| BC140977.1 | - | 10 - 11 | 1086 | 9055589 | 112 | |
More Information
Set Membership: |
|

Xpnpep2
Genomic Map