Assay Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133216.3 | NP_573479.3 | 9 - 10 | 938 | 138 | |
| GenBank mRNA | AF363970.1 | - | 9 - 10 | 878 | 138 | |
| AK052094.1 | - | 1 - 2 | 80 | 138 | ||
| AK146033.1 | - | 9 - 10 | 918 | 138 | ||
| AK149676.1 | - | 9 - 10 | 877 | 138 | ||
| AK158597.1 | - | 7 - 8 | 896 | 138 | ||
| AK166715.1 | - | 9 - 10 | 838 | 138 | ||
| AK169024.1 | - | 8 - 9 | 764 | 138 | ||
| BC065174.1 | - | 9 - 10 | 826 | 6415314 | 138 | |
More Information
Set Membership: |
|

Xpnpep1
Genomic Map