Assay Details
| Gene Symbol: Myoz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133363.3 | NP_579941.2 | 6 - 7 | 591 | 109 | |
| GenBank mRNA | AF480442.1 | - | 6 - 7 | 669 | 109 | |
| BC104046.2 | - | 6 - 7 | 591 | 40040057 | 109 | |
| BC104047.1 | - | 6 - 7 | 591 | 40040058 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752325 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752327 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752322 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752324 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752323 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752321 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752326 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963239 | BC104046.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963239 | BC104047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963239 | NM_133363.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963239 | AF480442.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2217438 | AK014729.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118252 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146057 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100102 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963239 | BC104046.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963239 | BC104047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963239 | NM_133363.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963239 | AF480442.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2217438 | AK014729.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31752299 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118252 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100102 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146057 | - | - |
| Whole Mouse Genome G4122A | A_51_P337974 | AF480442.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103927 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109136 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100102 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146057 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118252 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13757655 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13757656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13757654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13849807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13849803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13849811 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752326 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752321 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752327 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752325 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752323 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752324 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31752322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31752191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13757662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13897602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13849806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14124662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13597582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13897600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14124663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13723063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13597585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14004226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13597590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31752251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10459079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459083 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450365_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430738_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10459075 | - | - |
| Mouse Genome 430 2.0 Array | 1450365_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450365_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170947_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170947_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800010207.mm.1 | NM_133363.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010207.mm.1 | NM_133363.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010207.mm.1 | TC1800001401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800010205.mm.1 | NM_133363.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010205.mm.1 | NM_133363.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010205.mm.1 | TC1800001401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800010204.mm.1 | NM_133363.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010204.mm.1 | NM_133363.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800010204.mm.1 | TC1800001401.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450365_PM_at | - | - |
| Mouse Expression Set 430 | 1450365_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450365_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450365_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450365_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450365_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493798 | - | - |
| ABI Mouse Genome Survey Microarray | 526851 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1990138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226637 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226637 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3081836 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3011346 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3011346 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3081836 | - | - |

Myoz3
Genomic Map