Assay Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133946.2 | NP_598707.2 | 7 - 8 | 2674 | 84 | |
| GenBank mRNA | AK039531.1 | - | 4 - 5 | 1958 | 84 | |
| AY355343.1 | - | 4 - 5 | 1612 | 84 | ||
| BC013519.1 | - | 4 - 5 | 1581 | 84 | ||
| BC031139.1 | - | 4 - 5 | 1666 | 4972485 | 84 | |
| FJ222825.1 | - | 7 - 8 | 2620 | 84 | ||
| JF810536.1 | - | 6 - 7 | 3059 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
response to bacterium
regulation of autophagy
negative regulation of toll-like receptor signaling pathway
wound healing
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAPK cascade
regulation of inflammatory response
negative regulation of immune response
regulation of mucus secretion
nucleotide binding
vasopressin receptor activity
ATP binding
peptide binding
G-protein coupled receptor signaling pathway
response to bacterium
regulation of autophagy
negative regulation of toll-like receptor signaling pathway
wound healing
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAPK cascade
regulation of inflammatory response
negative regulation of immune response
regulation of mucus secretion
nucleotide binding
vasopressin receptor activity
ATP binding
peptide binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585683 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585680 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585677 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585678 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585682 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111021 | FJ222825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111021 | BC013519.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111021 | BC031139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111021 | AK039531.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111021 | AY355343.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71928 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149197 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111021 | FJ222825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111021 | BC013519.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111021 | BC031139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111021 | AK039531.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111021 | AY355343.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149197 | - | - |
| Whole Mouse Genome G4122A | A_51_P362818 | BC013519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362818 | BC031139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362818 | AK039531.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362818 | AY355343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362818 | NM_001081389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98531 | BC031139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98531 | AY355343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P98531 | NM_001081389.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111021 | FJ222825.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111021 | BC013519.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111021 | BC031139.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111021 | AK039531.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111021 | AY355343.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112389 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585680 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585683 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585677 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585682 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25585678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06208844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06432331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06503522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06432335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06267956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06267962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06267944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05965216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06267966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05965214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06435613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05965220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585636 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558750 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558748 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427369_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558742 | - | - |
| Mouse Genome 430 2.0 Array | 1427369_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427369_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101613_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101613_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017873.mm.1 | TC0700002032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017873.mm.1 | NM_133946.2 | Refseq |
| Mouse Expression Set 430 | 1427369_at | - | - |
| Mouse Expression Set 430 | 1427369_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427369_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427369_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427369_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427369_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 578850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2918203 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2918203 | - | - |

Nlrp6
Genomic Map