Assay Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134151.4 | NP_598912.4 | 2 - 3 | 425 | 92 | |
| GenBank mRNA | AK028785.1 | - | 2 - 3 | 287 | 92 | |
| AK043837.1 | - | 2 - 3 | 349 | 92 | ||
| AK076634.1 | - | 2 - 3 | 270 | 92 | ||
| AK139579.1 | - | 2 - 3 | 270 | 92 | ||
| AK145356.1 | - | 2 - 3 | 318 | 92 | ||
| AK151880.1 | - | 2 - 3 | 277 | 92 | ||
| AK169708.1 | - | 2 - 3 | 201 | 92 | ||
| AK195910.1 | - | 1 - 2 | 58 | 92 | ||
| AK200644.1 | - | 1 - 2 | 58 | 92 | ||
| AK205652.1 | - | 1 - 2 | 58 | 92 | ||
| BC013552.1 | - | 2 - 3 | 356 | 4236514 | 92 | |
| BC022143.1 | - | 2 - 3 | 320 | 5101324 | 92 | |
| BC026615.1 | - | 2 - 3 | 332 | 4976999 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yars
Genomic Map