Assay Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134151.4 | NP_598912.4 | 7 - 8 | 1041 | 68 | |
| GenBank mRNA | AK028785.1 | - | 7 - 8 | 903 | 68 | |
| AK043837.1 | - | 7 - 8 | 965 | 68 | ||
| AK076634.1 | - | 7 - 8 | 886 | 68 | ||
| AK139579.1 | - | 7 - 8 | 886 | 68 | ||
| AK145356.1 | - | 7 - 8 | 934 | 68 | ||
| AK151880.1 | - | 7 - 8 | 893 | 68 | ||
| AK169708.1 | - | 7 - 8 | 817 | 68 | ||
| AK219154.1 | - | 1 - 2 | 65 | 68 | ||
| BC013552.1 | - | 7 - 8 | 972 | 4236514 | 68 | |
| BC022143.1 | - | 7 - 8 | 936 | 5101324 | 68 | |
| BC026615.1 | - | 7 - 8 | 948 | 4976999 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yars
Genomic Map