Assay Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134157.2 | NP_598918.1 | 8 - 9 | 849 | 77 | |
| GenBank mRNA | AF435091.1 | - | 8 - 9 | 849 | 77 | |
| AK052604.1 | - | 8 - 9 | 820 | 77 | ||
| AK052707.1 | - | 8 - 9 | 811 | 77 | ||
| AK078810.1 | - | 8 - 9 | 877 | 77 | ||
| AK085549.1 | - | 8 - 9 | 850 | 77 | ||
| AK168980.1 | - | 8 - 9 | 877 | 77 | ||
| BC004789.1 | - | 5 - 6 | 443 | 77 | ||
| BC017127.1 | - | 8 - 9 | 834 | 4489144 | 77 | |
| BC062202.1 | - | 8 - 9 | 808 | 2650839 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b1
Genomic Map