Assay Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134157.2 | NP_598918.1 | 9 - 10 | 982 | 72 | |
| GenBank mRNA | AF435091.1 | - | 9 - 10 | 982 | 72 | |
| AK052604.1 | - | 9 - 10 | 953 | 72 | ||
| AK052707.1 | - | 9 - 10 | 944 | 72 | ||
| AK078810.1 | - | 9 - 10 | 1010 | 72 | ||
| AK085549.1 | - | 9 - 10 | 983 | 72 | ||
| AK168980.1 | - | 9 - 10 | 1010 | 72 | ||
| BC004789.1 | - | 6 - 7 | 576 | 72 | ||
| BC017127.1 | - | 9 - 10 | 967 | 4489144 | 72 | |
| BC062202.1 | - | 9 - 10 | 941 | 2650839 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b1
Genomic Map