Assay Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144783.2 | NP_659032.3 | 2 - 3 | 1042 | 101 | |
| GenBank mRNA | AK052767.1 | - | 1 - 2 | 2267 | 101 | |
| DQ537939.1 | - | 2 - 3 | 379 | 101 | ||
| M55512.1 | - | 2 - 3 | 1047 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wt1
Genomic Map