Assay Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144783.2 | NP_659032.3 | 4 - 5 | 1222 | 76 | |
| GenBank mRNA | DQ537939.1 | - | 4 - 5 | 559 | 76 | |
| M55512.1 | - | 4 - 5 | 1227 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wt1
Genomic Map