Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100031437.mm.1 |
NM_144817.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031437.mm.1 |
NM_144817.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031437.mm.1 |
TC0100003840.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031439.mm.1 |
NM_144817.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031439.mm.1 |
NM_144817.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031439.mm.1 |
TC0100003840.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031438.mm.1 |
NM_144817.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031438.mm.1 |
NM_144817.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031438.mm.1 |
TC0100003840.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031436.mm.1 |
NM_144817.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031436.mm.1 |
NM_144817.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031436.mm.1 |
TC0100003840.mm.1 |
MTA TC |