Assay Details
| Gene Symbol: Cbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144855.3 | NP_659104.1 | 16 - 17 | 1836 | 92 | |
| GenBank mRNA | AK085692.1 | - | 16 - 17 | 1804 | 93 | |
| BC013480.1 | - | 16 - 17 | 1772 | 4239757 | 92 | |
| BC022144.1 | - | 16 - 17 | 1797 | 92 | ||
More Information
Set Membership: |
|

Cbs
Genomic Map