Assay Details
| Gene Symbol: Lppr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290299.1 | NP_001277228.1 | 6 - 7 | 1002 | 57 | |
| NM_144935.1 | NP_659184.1 | 5 - 6 | 974 | 57 | ||
| GenBank mRNA | AK082453.1 | - | 6 - 7 | 965 | 57 | |
| AY438023.1 | - | 5 - 6 | 974 | 57 | ||
| BC018242.1 | - | 5 - 6 | 974 | 4503683 | 57 | |
| BC023082.1 | - | 6 - 7 | 979 | 5357160 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lppr2
Genomic Map