Assay Details
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144935.1 | NP_659184.1 | 7 - 8 | 1251 | 68 | |
GenBank mRNA | AY438023.1 | - | 7 - 8 | 1251 | 68 | |
BC018242.1 | - | 7 - 8 | 1251 | 4503683 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
