Assay Details
| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145970.1 | NP_001139442.1 | 9 - 10 | 1751 | 96 | |
| NM_144941.3 | NP_659190.3 | 9 - 10 | 1967 | 96 | ||
| GenBank mRNA | AK122462.1 | - | 3 - 4 | 3390 | 96 | |
| AK151613.1 | - | 7 - 8 | 1482 | 96 | ||
| BC019977.1 | - | 4 - 5 | 943 | 4187506 | 96 | |
| BC023677.1 | - | 9 - 10 | 1942 | 5354497 | 96 | |
| BC023774.1 | - | 9 - 10 | 1748 | 96 | ||
| CK617787.1 | - | 2 - 3 | 422 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d1
Genomic Map