Assay Details
| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145970.1 | NP_001139442.1 | 13 - 14 | 2239 | 80 | |
| NM_144941.3 | NP_659190.3 | 13 - 14 | 2455 | 80 | ||
| GenBank mRNA | AK151613.1 | - | 11 - 12 | 1970 | 80 | |
| BC016081.1 | - | 4 - 5 | 491 | 80 | ||
| BC019977.1 | - | 8 - 9 | 1431 | 4187506 | 80 | |
| BC023677.1 | - | 13 - 14 | 2430 | 5354497 | 80 | |
| BC023774.1 | - | 13 - 14 | 2236 | 80 | ||
| BM944422.1 | - | 4 - 5 | 345 | 80 | ||
More Information
Set Membership: |
|

Map7d1
Genomic Map